missing translation for 'onlineSavingsMsg'
Learn More

Sodium ethoxide, 21% in ethanol, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10568505
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-52-6, 64-17-5
Molecular Formula C2H5NaO
Molecular Weight (g/mol) 68.04
MDL Number MFCD00012417
InChI Key QDRKDTQENPPHOJ-UHFFFAOYSA-N
Synonym sodium ethoxide, sodium ethylate, sodium ethanolate, sodiumethoxide, ethoxysodium, ethanol, sodium salt, caustic alcohol, naoet, etona, ethanol, sodium salt 1:1
PubChem CID 2723922
ChEBI CHEBI:52096
IUPAC Name sodium;ethanolate
SMILES CC[O-].[Na+]

Specifications

Boiling Point 91.0°C
Quantity 100 mL
Linear Formula C2H5ONa
Flash Point 25°C
Merck Index 15,8749
Solubility Information Solubility in water: miscible (hydrolyse)
Formula Weight 68.04
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.385
Specific Gravity 0.868
Density 0.8680g/mL
Chemical Name or Material Sodium ethoxide, 21% in ethanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium ethoxide, 21% in ethanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

For Manufacturing and Laboratory Use. Not for beverage use

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'