Learn More
Sodium ethoxide, 96%, pure, Thermo Scientific Chemicals
£34.40 - £180.00
Chemical Identifiers
CAS | 141-52-6 |
---|---|
Molecular Formula | C2H5NaO |
Molecular Weight (g/mol) | 68.04 |
MDL Number | MFCD00012417 |
InChI Key | QDRKDTQENPPHOJ-UHFFFAOYSA-N |
Synonym | sodium ethoxide, sodium ethylate, sodium ethanolate, sodiumethoxide, ethoxysodium, ethanol, sodium salt, caustic alcohol, naoet, etona, ethanol, sodium salt 1:1 |
PubChem CID | 2723922 |
ChEBI | CHEBI:52096 |
IUPAC Name | sodium;ethanolate |
SMILES | CC[O-].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10215600
|
Thermo Scientific Acros
168592500 |
250 g | Glass bottle |
£34.40
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10225600
|
Thermo Scientific Acros
168595000 |
500 g | Glass bottle |
£67.30
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10784104
|
Thermo Scientific Acros
168590025 |
2.5 kg | Plastic Drum |
£180.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
141-52-6 | |
68.04 | |
QDRKDTQENPPHOJ-UHFFFAOYSA-N | |
2723922 | |
sodium;ethanolate |
C2H5NaO | |
MFCD00012417 | |
sodium ethoxide, sodium ethylate, sodium ethanolate, sodiumethoxide, ethoxysodium, ethanol, sodium salt, caustic alcohol, naoet, etona, ethanol, sodium salt 1:1 | |
CHEBI:52096 | |
CC[O-].[Na+] |
Specifications
141-52-6 | |
0.868 | |
C2H5NaO | |
MFCD00012417 | |
01,1065; 03,265; 04,451; 06,540 | |
sodium ethoxide, sodium ethylate, sodium ethanolate, sodiumethoxide, ethoxysodium, ethanol, sodium salt, caustic alcohol, naoet, etona, ethanol, sodium salt 1:1 | |
QDRKDTQENPPHOJ-UHFFFAOYSA-N | |
sodium;ethanolate | |
2723922 | |
68.04 | |
Pure |
260.0°C | |
95% min. (NaOC2H5) | |
C2H5ONa | |
01, 311 | |
15, 8749 | |
Solubility in water: reacts. Other solubilities: soluble in absolute alcohol without decomposition | |
CC[O-].[Na+] | |
68.04 | |
CHEBI:52096 | |
96% | |
Sodium ethoxide, 96% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Self-heating; may catch fire.
Flammable solid.
Reacts violently with water.
GHS P Statement
Keep cool.
Protect from sunlight.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 205-487-5
missing translation for 'tsca' : TSCA
- Sodium ethoxide
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 1
- Self-heating substance Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H251-Self-heating: may catch fire.
- H228-Flammable solid.
- EUH014-Reacts violently with water.
Precautionary Statement
- P235+P410-Keep cool. Protect from sunlight.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only