missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium fluoride, 97%, extra pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
Specifications
Boiling Point | 1700.0°C |
Melting Point | 993.0°C |
Color | White |
Physical Form | Crystals or Powder |
Quantity | 5 kg |
Assay Percent Range | 97% |
Linear Formula | NaF |
Fieser | 01,1073; 02,382 |
Merck Index | 15, 8754 |
Solubility Information | Solubility aq. soln.: 40 g/L (25°C). Other solubilities: slightly soluble in alcohol |
Show More |
Safety and Handling
Product Identifier
- Sodium fluoride
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H301-Toxic if swallowed.
- EUH032-Contact with acids liberates very toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'