missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Fluoride, 99.0+%, Certified AR for Analysis, Fisher Chemical™
£121.00 - £344.00
Chemical Identifiers
CAS | 7681-49-4 |
---|---|
Molecular Formula | FNa |
Molecular Weight (g/mol) | 41.99 |
MDL Number | MFCD00003524 |
InChI Key | PUZPDOWCWNUUKD-UHFFFAOYSA-M |
Synonym | sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril |
PubChem CID | 5235 |
ChEBI | CHEBI:28741 |
SMILES | [F-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10244540
|
Fisher Chemical
S/3920/50 |
250 g |
£121.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10376240
|
Fisher Chemical
S/3920/53 |
500 g |
£219.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10386240
|
Fisher Chemical
S/3920/60 |
1 kg |
£325.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12676767
|
Fisher Chemical
S/3920/63 |
3 kg | N/A | ||||||
Description
Sodium Fluoride, 99.0+%, Certified AR for Analysis
Chemical Identifiers
7681-49-4 | |
41.99 | |
PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
5235 | |
[F-].[Na+] |
FNa | |
MFCD00003524 | |
sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril | |
CHEBI:28741 |
Specifications
1700°C | |
7681-49-4 | |
7.4 | |
FNa | |
sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril | |
[F-].[Na+] | |
41.99 | |
CHEBI:28741 | |
Plastic powder jar |
993°C | |
White | |
Powder/Solid | |
MFCD00003524 | |
PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
sodium;fluoride | |
5235 | |
41.98g/mol | |
1mmHg 1077 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Sodium fluoride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H301-Toxic if swallowed.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /A§
- P302+P352-IF ON SKIN: Wash with plenty of water/A§.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.