Learn More
Sodium fluoride, 99+%, for analysis, Thermo Scientific Chemicals
£15.48 - £254.27
Chemical Identifiers
CAS | 7681-49-4 |
---|---|
Molecular Formula | FNa |
Molecular Weight (g/mol) | 41.99 |
MDL Number | MFCD00003524 |
InChI Key | PUZPDOWCWNUUKD-UHFFFAOYSA-M |
Synonym | sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril |
PubChem CID | 5235 |
ChEBI | CHEBI:28741 |
IUPAC Name | sodium fluoride |
SMILES | [F-].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10296970
|
Thermo Scientific Acros
201290250 |
25 g | Plastic bottle |
£15.48
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10113980
|
Thermo Scientific Acros
201295000 |
500 g | Plastic bottle |
£62.61
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10761994
|
Thermo Scientific Acros
201290025 |
2.5 kg | Plastic bottle |
£254.27
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
7681-49-4 | |
41.99 | |
PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
5235 | |
sodium fluoride |
FNa | |
MFCD00003524 | |
sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril | |
CHEBI:28741 | |
[F-].[Na+] |
Specifications
1700.0°C | |
7681-49-4 | |
Powder | |
FNa | |
MFCD00003524 | |
15, 8754 | |
Solubility in water: 40g/L (25°C). Other solubilities: slightly soluble in alcohol | |
[F-].[Na+] | |
41.99 | |
CHEBI:28741 | |
≥99% | |
0.2% max. (150°C) |
993.0°C | |
White | |
99+% | |
NaF | |
01,1073; 02,382 | |
sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril | |
PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
sodium fluoride | |
5235 | |
41.98 | |
Analytical | |
Sodium fluoride, p.a. |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Contact with acids liberates very toxic gas.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-667-8
missing translation for 'rtecsNumber' : WB0350000
missing translation for 'tsca' : TSCA
- Sodium fluoride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only