Learn More
Sodium fluoride, ACS, 99% min, Thermo Scientific Chemicals
Steel Degreasing Agent, in Tin Electroplating, Flux
£39.40 - £121.00
Chemical Identifiers
CAS | 7681-49-4 |
---|---|
Molecular Formula | FNa |
Molecular Weight (g/mol) | 41.99 |
MDL Number | MFCD00003524 |
InChI Key | PUZPDOWCWNUUKD-UHFFFAOYSA-M |
Synonym | sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril |
PubChem CID | 5235 |
ChEBI | CHEBI:28741 |
IUPAC Name | sodium fluoride |
SMILES | [F-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11307287
|
Thermo Scientific Alfa Aesar
011561.30 |
250 g |
£39.40
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11317287
|
Thermo Scientific Alfa Aesar
011561.A1 |
1 kg |
£121.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a steel degreasing agent and in tin electroplating, flux, heat treating salt compositions, fluoridation of drinking water, removing HF from exhaust systems to reduce air pollution, and frosting glasses. It serves as a source of fluoride ion in several applications including it use in synthetic chemistry, for example in Finkelstein reaction for preparating fluoro compounds, desilylation in sensitive organic compounds, metallurgy, and reaction with electrophilic chlorides. It has medical and other applications including complexing of magnesium ions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7681-49-4 | |
41.99 | |
PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
5235 | |
sodium fluoride |
FNa | |
MFCD00003524 | |
sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril | |
CHEBI:28741 | |
[F-].[Na+] |
Specifications
1704°C | |
7681-49-4 | |
Powder | |
FNa | |
sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium fluoride, antibulit, floridine, fluonatril | |
PUZPDOWCWNUUKD-UHFFFAOYSA-M | |
sodium fluoride | |
5235 | |
41.99 | |
Odorless | |
White powder | |
2.78 g/cm3 at 20°C |
993°C | |
White | |
99% min. | |
MFCD00003524 | |
Slightly Soluble in water. Insoluble in alcohol | |
[F-].[Na+] | |
41.99 | |
CHEBI:28741 | |
ACS | |
1.336 | |
1.3 hPa (1mm Hg) | |
Sodium fluoride, For ACS analysis |
Safety and Handling
P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H301-H315-H319
missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: III
missing translation for 'einecsNumber' : 231-667-8
missing translation for 'rtecsNumber' : WB0350000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Sodium fluoride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : sodium fluoride
RUO – Research Use Only