missing translation for 'onlineSavingsMsg'
Learn More
Sodium formate, 98% Product Code.: 11423408

Sodium formate, 98%

Product Code. 11423408
1000 g, 1000g
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
This item is not returnable. View return policy

Product Code. 11423408

Brand: Thermo Scientific Alfa Aesar A17813.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 141-53-7 | CHNaO2 | 68.007 g/mol

Sodium formate is used in analytical chemistry to precipitate noble metals. It is used for tanning leather, for dyeing and printing fabrics. It acts as an electroplating agent, an acidulant in the textile industry, a reducing agent, a mordant, a complexing agent. Further, it is used in photographic fixing baths. In addition, it is used as a buffering agent for strong mineral acids and food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium formate is used in analytical chemistry to precipitate noble metals. It is used for tanning leather, for dyeing and printing fabrics. It acts as an electroplating agent, an acidulant in the textile industry, a reducing agent, a mordant, a complexing agent. Further, it is used in photographic fixing baths. In addition, it is used as a buffering agent for strong mineral acids and food additive.

Solubility
Soluble in water, alcohol, glycerol and formic acid. Insoluble in ether.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong acids. Moisture sensitive
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-53-7
Molecular Formula CHNaO2
Molecular Weight (g/mol) 68.007
MDL Number MFCD00013101
InChI Key HLBBKKJFGFRGMU-UHFFFAOYSA-M
Synonym sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744
PubChem CID 2723810
ChEBI CHEBI:62965
IUPAC Name sodium;formate
SMILES C(=O)[O-].[Na+]

Specifications

Melting Point 259°C to 262°C
Density 1.92
Quantity 1000 g
Beilstein 3595134
Sensitivity Hygroscopic
Merck Index 14,8621
Solubility Information Soluble in water,alcohol,glycerol and formic acid. Insoluble in ether.
Formula Weight 68.01
Percent Purity 98%
Chemical Name or Material Sodium formate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number10238780 Certificate TypeCertificate of Analysis Date24/02/2023 Product CodeA17813.30,A17813.0B,A17813.0I
Lot Number10245534 Certificate TypeCertificate of Analysis Date30/01/2023 Product CodeA17813.30,A17813.0B,A17813.0I
Lot Number10244812 Certificate TypeCertificate of Analysis Date30/11/2022 Product CodeA17813.30,A17813.0B,A17813.0I
Lot Number10240356 Certificate TypeCertificate of Analysis Date29/04/2022 Product CodeA17813.30,A17813.0B,A17813.0I
Lot Number10237557 Certificate TypeCertificate of Analysis Date12/04/2022 Product CodeA17813.30,A17813.0B,A17813.0I

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium formate, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.