missing translation for 'onlineSavingsMsg'
Learn More
Sodium formate, ACS reagent Product Code.: 12386263

Sodium formate, ACS reagent

Product Code. 12386263 Shop All Thermo Scientific Chemicals Products
100 g, Plastic bottle, 100g
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 12386263

Brand: Thermo Scientific Acros 446951000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 141-53-7 | CHNaO2 | 68.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-53-7
Molecular Formula CHNaO2
Molecular Weight (g/mol) 68.01
InChI Key HLBBKKJFGFRGMU-UHFFFAOYSA-M
Synonym sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744Show MoreShow Less
PubChem CID 2723810
ChEBI CHEBI:62965
IUPAC Name sodium;formate
SMILES C(=O)[O-].[Na+]

Specifications

Melting Point 258.0°C to 261.0°C
Boiling Point 360.0°C
Packaging Plastic bottle
Linear Formula HCO2Na
Quantity 100 g
Merck Index 158756
Solubility Information Solubility in water: 550g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol,insoluble in ether
Formula Weight 68.01
Grade ACS Reagent
Insoluble Matter 0.005% max
Chemical Name or Material Sodium formate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0461548 Certificate TypeCertificate of Analysis Date06/10/2025 Product Code446955000,446951000,446950025
Lot NumberA0461548 Certificate TypeCertificate of Analysis Date05/04/2024 Product Code446955000,446951000,446950025
Lot NumberA0399778 Certificate TypeCertificate of Analysis Date14/12/2022 Product Code446955000,446951000,446950025
Lot NumberA0413842 Certificate TypeCertificate of Analysis Date13/12/2022 Product Code446955000,446951000,446950025
Lot NumberA0447311 Certificate TypeCertificate of Analysis Date11/11/2022 Product Code446955000,446951000,446950025

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium formate, ACS reagent >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.