missing translation for 'onlineSavingsMsg'
Learn More

Sodium Formate, Honeywell Fluka™

Product Code. 15651580
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

for HPLC,≥99.0% (NT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-53-7
Molecular Formula CHNaO2
Molecular Weight (g/mol) 68.007
MDL Number MFCD00013101
InChI Key HLBBKKJFGFRGMU-UHFFFAOYSA-M
Synonym sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744
PubChem CID 2723810
ChEBI CHEBI:62965
IUPAC Name sodium;formate
SMILES C(=O)[O-].[Na+]

Specifications

Quantity 50 g
Linear Formula HCOONa
UN Number NONH for all modes of transport
Beilstein 3595134
Solubility Information Soluble in water 0.1g/mL at 25°C
Formula Weight 68.01g/mol
Percent Purity ≥99% (NT)
Chemical Name or Material Sodium Formate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'