missing translation for 'onlineSavingsMsg'
Learn More

Sodium glycolate, 97%

Product Code. 11459463
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11459463

Brand: Thermo Scientific Alfa Aesar A12341.0E

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2836-32-0 | C2H3NaO3 | 98.033 g/mol

Sodium glycolate is used as a disintegrant, suspending agent, gelling agent, buffering agent and in pharmaceutical grade for tablets and capsules. Further, it is used in cosmetics and personal care products primarily as an exfoliant, as pH adjusters, skin-conditioning agents and as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium glycolate is used as a disintegrant, suspending agent, gelling agent, buffering agent and in pharmaceutical grade for tablets and capsules. Further, it is used in cosmetics and personal care products primarily as an exfoliant, as pH adjusters, skin-conditioning agents and as a flavoring agent.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2836-32-0
Molecular Formula C2H3NaO3
Molecular Weight (g/mol) 98.033
MDL Number MFCD00065358
InChI Key VILMUCRZVVVJCA-UHFFFAOYSA-M
Synonym sodium glycolate, sodium hydroxyacetate, sodium 2-hydroxyacetate, sodium lycolate, monosodium glycolate, glycolic acid sodium salt, natriumglykolat, sodium glycollate, acetic acid, hydroxy-, monosodium salt, glycolic acid, monosodium salt
PubChem CID 517347
IUPAC Name sodium;2-hydroxyacetate
SMILES C(C(=O)[O-])O.[Na+]

Specifications

Melting Point 210°C to 218°C
Assay Percent Range 97%
Linear Formula HOCH2CO2Na
Quantity 2500 g
Solubility Information Soluble in water.
Formula Weight 98.03
Percent Purity 97%
Physical Form Powder or Lumps
Chemical Name or Material Sodium glycolate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.