missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hexafluoroantimonate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
Specifications
Color | White |
Physical Form | Fine Crystalline Powder |
Quantity | 25 g |
Assay Percent Range | 97.5% min |
Linear Formula | NaSbF6 |
Solubility Information | Solubility in water: very soluble |
Formula Weight | 258.72 |
Percent Purity | 98% |
Packaging | Plastic bottle |
Chemical Name or Material | Sodium hexafluoroantimonate, 98% |
Safety and Handling
Product Identifier
- Sodium hexafluoroantimonate
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- H411-Toxic to aquatic life with long lasting effects.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'