Learn More
Sodium hexafluoroantimonate(V), 98%, Thermo Scientific Chemicals
£35.28 - £393.96
Chemical Identifiers
CAS | 16925-25-0 |
---|---|
Molecular Formula | F6NaSb |
Molecular Weight (g/mol) | 258.74 |
MDL Number | MFCD00003482 |
InChI Key | HKLMYZVMEYYVBS-UHFFFAOYSA-H |
Synonym | sodium hexafluoroantimonate, sodium hexafluoroantimonate v, sodium hexafluorostibanuide, sodium hexafluoro antimonate, antimony sodium fluoride, f6sb.na, nasbf6, acmc-20ajo7, sodium hexafluorostiboranuide, hexafluorostibine, sodium saltShow More |
PubChem CID | 16689647 |
IUPAC Name | sodium;hexafluoroantimony(1-) |
SMILES | F[Sb-](F)(F)(F)(F)F.[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11499017
|
Thermo Scientific Alfa Aesar
B20604.14 |
25 g |
£35.28
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11409027
|
Thermo Scientific Alfa Aesar
B20604.22 |
100 g |
£96.48
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11419027
|
Thermo Scientific Alfa Aesar
B20604.36 |
500 g |
£393.96
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16925-25-0 | |
258.74 | |
HKLMYZVMEYYVBS-UHFFFAOYSA-H | |
16689647 | |
F[Sb-](F)(F)(F)(F)F.[Na+] |
F6NaSb | |
MFCD00003482 | |
sodium hexafluoroantimonate, sodium hexafluoroantimonate v, sodium hexafluorostibanuide, sodium hexafluoro antimonate, antimony sodium fluoride, f6sb.na, nasbf6, acmc-20ajo7, sodium hexafluorostiboranuide, hexafluorostibine, sodium saltShow More | |
sodium;hexafluoroantimony(1-) |
Specifications
16925-25-0 | |
MFCD00003482 | |
sodium hexafluoroantimonate, sodium hexafluoroantimonate v, sodium hexafluorostibanuide, sodium hexafluoro antimonate, antimony sodium fluoride, f6sb.na, nasbf6, acmc-20ajo7, sodium hexafluorostiboranuide, hexafluorostibine, sodium salt | |
F[Sb-](F)(F)(F)(F)F.[Na+] | |
258.74 | |
258.73 | |
Odorless | |
Sodium hexafluoroantimonate(V) |
F6NaSb | |
UN1549 | |
HKLMYZVMEYYVBS-UHFFFAOYSA-H | |
sodium;hexafluoroantimony(1-) | |
16689647 | |
98% | |
3.38 |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
missing translation for 'einecsNumber' : 240-989-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.