missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexafluorophosphate, 99+%, Thermo Scientific Chemicals

Product Code. 11342158
Click to view available options
Quantity:
25 g
100 g

Sodium hexafluorophosphate is used as an ion pairing agent for the determination of cobalt(II) in water samples. It is used as an important raw material in the preparation of 2-chloro-1,3-bis(dimethylamino)trimethinium hexafluorophosphate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21324-39-0
Molecular Formula F6NaP
Molecular Weight (g/mol) 167.95
MDL Number MFCD00011122
InChI Key KMADQUOCJBLXRP-UHFFFAOYSA-N
Synonym sodium hexafluorophosphate, sodium hexa-fluoro phosphate, sodium hexafluorophosphate v, napf6, sodium hexaflurophosphate, acmc-1cao2, sodium hexafluoro-, e5-phosphanuide, sodium hexafluoro-$l^ 5-phosphanuide, phosphate 1-, hexafluoro-, sodium 1:1
PubChem CID 5147921
IUPAC Name sodium;hexafluorophosphate
SMILES [Na+].F[P-](F)(F)(F)(F)F

Specifications

Melting Point >200°C
Color White
Physical Form Crystals
Quantity 25 g
Assay Percent Range >99%
Solubility Information Soluble in water.
Formula Weight 167.95
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystalline
Density 2.37
Chemical Name or Material Sodium hexafluorophosphate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'