missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hexafluorophosphate, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Sodium hexafluorophosphate is used as an ion pairing agent for the determination of cobalt(II) in water samples. It is used as an important raw material in the preparation of 2-chloro-1,3-bis(dimethylamino)trimethinium hexafluorophosphate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | >200°C |
Color | White |
Physical Form | Crystals |
Quantity | 100 g |
Assay Percent Range | >99% |
Solubility Information | Soluble in water. |
Formula Weight | 167.95 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Appearance | White crystalline |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'