Learn More
Sodium hexahydroxyplatinate(IV), 99.9% (metals basis), Thermo Scientific Chemicals
£243.00 - £977.00
Chemical Identifiers
CAS | 12325-31-4 |
---|---|
Molecular Formula | H6Na2O6Pt |
Molecular Weight (g/mol) | 343.11 |
MDL Number | MFCD00014236 |
InChI Key | LDMNYTKHBHFXNG-UHFFFAOYSA-H |
IUPAC Name | platinum(4+) disodium hexahydroxide |
SMILES | [OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Na+].[Na+].[Pt+4] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11327477
|
Thermo Scientific Alfa Aesar
012620.03 |
1 g |
£243.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11317477
|
Thermo Scientific Alfa Aesar
012620.06 |
5 g |
£977.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12325-31-4 | |
343.11 | |
LDMNYTKHBHFXNG-UHFFFAOYSA-H | |
[OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Na+].[Na+].[Pt+4] |
H6Na2O6Pt | |
MFCD00014236 | |
platinum(4+) disodium hexahydroxide |
Specifications
12325-31-4 | |
Yellow to Brown | |
Na2Pt(OH)6 | |
LDMNYTKHBHFXNG-UHFFFAOYSA-H | |
platinum(4+) disodium hexahydroxide | |
343.1 | |
Crystalline | |
Sodium hexahydroxyplatinate(IV) |
150°C (decomposition) | |
H6Na2O6Pt | |
MFCD00014236 | |
[OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Na+].[Na+].[Pt+4] | |
343.11 | |
99.9% | |
(metals basis) |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'einecsNumber' : 235-590-0
missing translation for 'rtecsNumber' : TP1756000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only