missing translation for 'onlineSavingsMsg'
Learn More
Sodium Hexametaphosphate, SLR, Powder, Mainly (NaPO3)6, Fisher Chemical™ Product Code.: 10204640

Sodium Hexametaphosphate, SLR, Powder, Mainly (NaPO3)6, Fisher Chemical™

Product Code. 10204640
500 g
Click to view available options
Quantity:
500 g
1 kg
This item is not returnable. View return policy

Product Code. 10204640

Brand: Fisher Chemical S/4160/53

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Na6O18P6, CAS Number-10124-56-8, 500g, White, 611.7708g/mol, 640 deg.C, 136045, Plastic powder jar, 6.0 to 7.7, Solid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10124-56-8
Molecular Formula Na6O18P6
Molecular Weight (g/mol) 611.77
MDL Number 136045

Specifications

Melting Point 640°C
Color White
pH 6.0 to 7.7
Physical Form Powder
Quantity 500 g
Formula Weight 611.7708g/mol
Packaging Plastic powder jar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Hexametaphosphate, SLR, Powder, Mainly (NaPO3)6, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.