missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hexametaphosphate, SLR, Powder, Mainly (NaPO3)6, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
Description
Na6O18P6, CAS Number-10124-56-8, 500g, White, 611.7708g/mol, 640 deg.C, 136045, Plastic powder jar, 6.0 to 7.7, Solid

Specifications
Specifications
Melting Point | 640°C |
Color | White |
pH | 6.0 to 7.7 |
Physical Form | Solid |
Quantity | 1 kg |
Formula Weight | 611.7708g/mol |
Packaging | Plastic powder jar |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
Chemicals Monthly Promotion
Take advantage of our exclusive monthly promotions on high-quality chemicals.Promo Code: 24198Expires: 2025-03-31
New Offer Save Up to 59% -
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Hexametaphosphate, SLR, Powder, Mainly (NaPO3)6, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction