missing translation for 'onlineSavingsMsg'
Learn More

Sodium hyaluronate, 95%, Thermo Scientific Chemicals

£63.42 - £644.21

Chemical Identifiers

CAS 9067-32-7
Molecular Formula (C14H20NO11Na)n
Molecular Weight (g/mol) 417.30
MDL Number MFCD00875848
InChI Key YWIVKILSMZOHHF-QJZPQSOGSA-N
Synonym hyaluronate tetrasaccharide, d0e9sz, 6-3-acetamido-2-6-3-acetamido-2,5-dihydroxy-6-hydroxymethyl oxan-4-yl oxy-2-carboxy-4,5-dihydroxyoxan-3-yl oxy-5-hydroxy-6-hydroxymethyl oxan-4-yl oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
IUPAC Name Sodium hyaluronate
SMILES CC(=O)NC1C(OC2C(O)C(O)C(O-*)OC2C(=O)O[Na])OC(CO)C(O)C1O-*
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10491281
View Documents
Thermo Scientific Acros
251770010
1 g Glass bottle
£63.42
1g
Please to purchase this item. Need a web account? Register with us today!
10134623
View Documents Promotion Details
Thermo Scientific Acros
251770050
5 g Glass bottle
£200.00
5g
Please to purchase this item. Need a web account? Register with us today!
11771371
View Documents
Thermo Scientific Acros
251770250
25 g Glass bottle
£644.21
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9067-32-7
417.30
YWIVKILSMZOHHF-QJZPQSOGSA-N
Sodium hyaluronate
(C14H20NO11Na)n
MFCD00875848
hyaluronate tetrasaccharide, d0e9sz, 6-3-acetamido-2-6-3-acetamido-2,5-dihydroxy-6-hydroxymethyl oxan-4-yl oxy-2-carboxy-4,5-dihydroxyoxan-3-yl oxy-5-hydroxy-6-hydroxymethyl oxan-4-yl oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
CC(=O)NC1C(OC2C(O)C(O)C(O-*)OC2C(=O)O[Na])OC(CO)C(O)C1O-*

Specifications

9067-32-7
6.0 to 8.0 (0.5% aq. soln.)
Authentic
(C14H20NO11Na)n
MFCD00875848
hyaluronate tetrasaccharide, d0e9sz, 6-3-acetamido-2-6-3-acetamido-2,5-dihydroxy-6-hydroxymethyl oxan-4-yl oxy-2-carboxy-4,5-dihydroxyoxan-3-yl oxy-5-hydroxy-6-hydroxymethyl oxan-4-yl oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
YWIVKILSMZOHHF-QJZPQSOGSA-N
Sodium hyaluronate
95%
Sodium hyaluronate
White to Cream
10% max. (500mg, 105°C, 6 hrs)
94% min. (On dry substance)
(C14H20NO11Na)n
15,4796
Solubility in water: soluble.
CC(=O)NC1C(OC2C(O)C(O)C(O-*)OC2C(=O)O[Na])OC(CO)C(O)C1O-*
417.30
Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sodium hyaluronate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only