Learn More
Sodium hydrogen carbonate, ACS, 99.7-100.3%
Description
Sodium hydrogen carbonate is used as a laboratory chemical, a leavening agent, a neutralizer, as an active component in fire extinguishers, as a source of carbon dioxide and as a pH balancer in pools, as a cattle feed supplement and in spas and garden ponds. It is utilized for paint and corrosion removal, in baking powder, as a mild abrasive and a cleaning compound. In the field of medicine, it is used as an antacid to treat acid indigestion and heartburn.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Spezifikation
| Color | White |
| Physical Form | Granules |
| Quantity | 500 g |
| Assay Percent Range | 99.7-100.3% |
| Beilstein | 4153970 |
| Solubility Information | Soluble in water and methanol. Insoluble in ethanol. |
| IUPAC Name | sodium hydrogen carbonate |
| Formula Weight | 84.01 |
| Grade | ACS |
| Odor | Odorless |
| Mehr anzeigen |
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.