Learn More
Sodium hydrogen phosphate heptahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
£46.60 - £123.00
Chemical Identifiers
CAS | 7782-85-6 |
---|---|
Molecular Formula | H17Na2O11P |
Molecular Weight (g/mol) | 270.08 |
MDL Number | MFCD00149180 |
InChI Key | PYLIXCKOHOHGKQ-UHFFFAOYSA-N |
Synonym | sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate |
PubChem CID | 6096963 |
IUPAC Name | disodium;hydrogen phosphate;heptahydrate |
SMILES | O.O.O.O.O.O.O.[Na+].[Na+].OP(O)(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11386429
|
Thermo Scientific Alfa Aesar
011592.36 |
500 g |
£46.60
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11396429
|
Thermo Scientific Alfa Aesar
011592.A3 |
2 kg |
£123.00
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium hydrogen phosphate is widely used in detergents and cleaning agents. Combined with trisodium phosphate, it is employed in the food industry to adjust the pH and in water treatment to prevent calcium scale formation. It is utilized as a saline laxative to clean the bowel before a colonoscopy. It prevents the coagulation of condensed milk. It is also utilized as anti-caking additive in powdered products. It enhances the cook time and used as thickening agent in desserts and puddings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7782-85-6 | |
270.08 | |
PYLIXCKOHOHGKQ-UHFFFAOYSA-N | |
6096963 | |
O.O.O.O.O.O.O.[Na+].[Na+].OP(O)(O)=O |
H17Na2O11P | |
MFCD00149180 | |
sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate | |
disodium;hydrogen phosphate;heptahydrate |
Specifications
48.1°C-5H2O | |
White | |
98.0-102.0% | |
MFCD00149180 | |
Soluble in water and insoluble in ethanol. | |
O.O.O.O.O.O.O.[Na+].[Na+].OP(O)(O)=O | |
270.08 | |
268.07 (141.98 Anhydrous) | |
Odorless | |
1.7 g/cm3 at 20°C |
7782-85-6 | |
Crystalline | |
H17Na2O11P | |
sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate | |
PYLIXCKOHOHGKQ-UHFFFAOYSA-N | |
disodium;hydrogen phosphate;heptahydrate | |
6096963 | |
ACS | |
White crystals | |
Sodium hydrogen phosphate heptahydrate |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.
H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
missing translation for 'einecsNumber' : 231-448-7
missing translation for 'rtecsNumber' : WC4600000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only