missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydrogen Sulfate Monohydrate, Certified AR for Analysis, Fisher Chemical
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Specifications
Specifications
Melting Point | 58°C |
CAS | 10034-88-5 |
Color | White |
pH | 1 |
Physical Form | Solid |
Quantity | 500 g |
Molecular Formula | H3NaO5S |
MDL Number | MFCD00149210 |
Synonym | sodium bisulfate monohydrate, sodium hydrogen sulfate monohydrate, unii-3kls9y77yj, sulfuric acid, monosodium salt, hydrate, 3kls9y77yj, sodium hydrogensulfate hydrate, sulfuric acid, monosodium salt, monohydrate, sodium hydrate hydrogen sulfate, sodium hydrogen sulfate-1-hydrate, acmc-20ajox |
InChI Key | JXHZRQHZVYDRGX-UHFFFAOYSA-M |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Safety and Handling

Product Identifier
- Sodium bisulfate monohydrate
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Hydrogen Sulfate Monohydrate, Certified AR for Analysis, Fisher Chemical >
Spot an opportunity for improvement?Share a Content Correction