missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrogen sulfate monohydrate, Reagent Grade, Thermo Scientific Chemicals

£55.88 - £354.77

Chemical Identifiers

CAS 10034-88-5
Molecular Formula H3NaO5S
Molecular Weight (g/mol) 138.07
MDL Number MFCD00149210
InChI Key JXHZRQHZVYDRGX-UHFFFAOYSA-M
Synonym sodium bisulfate monohydrate, sodium hydrogen sulfate monohydrate, unii-3kls9y77yj, sulfuric acid, monosodium salt, hydrate, 3kls9y77yj, sodium hydrogensulfate hydrate, sulfuric acid, monosodium salt, monohydrate, sodium hydrate hydrogen sulfate, sodium hydrogen sulfate-1-hydrate, acmc-20ajox
PubChem CID 23673662
SMILES O.[Na+].OS([O-])(=O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11361339
View Documents
Thermo Scientific Alfa Aesar
013799.36
500 g
£55.88
500g
Please to purchase this item. Need a web account? Register with us today!
11381339
View Documents
Thermo Scientific Alfa Aesar
013799.A1
1 kg
£93.87
1kg
Please to purchase this item. Need a web account? Register with us today!
11371339
View Documents
Thermo Scientific Alfa Aesar
013799.A7
5 kg
£354.77
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sodium hydrogen sulfate monohydrate is used in metal finishing and cleaning products. It is used to lower the pH of water for an effective chlorination in swimming pools. It is also used as a urine acidifier to reduce urinary stones in cats. Further, it is used in controlling outbreaks of crown-of-thorns starfish. It is a primary ingredient used in jewelry making to clean the surfaces. In addition to this, it is used as a food additive and serves as a reagent for the decomposition of basic metal oxides by fusion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10034-88-5
138.07
JXHZRQHZVYDRGX-UHFFFAOYSA-M
23673662
H3NaO5S
MFCD00149210
sodium bisulfate monohydrate, sodium hydrogen sulfate monohydrate, unii-3kls9y77yj, sulfuric acid, monosodium salt, hydrate, 3kls9y77yj, sodium hydrogensulfate hydrate, sulfuric acid, monosodium salt, monohydrate, sodium hydrate hydrogen sulfate, sodium hydrogen sulfate-1-hydrate, acmc-20ajox
O.[Na+].OS([O-])(=O)=O

Specifications

∽183°C (decomposition)
White
H3NaO5S
sodium bisulfate monohydrate, sodium hydrogen sulfate monohydrate, unii-3kls9y77yj, sulfuric acid, monosodium salt, hydrate, 3kls9y77yj, sodium hydrogensulfate hydrate, sulfuric acid, monosodium salt, monohydrate, sodium hydrate hydrogen sulfate, sodium hydrogen sulfate-1-hydrate, acmc-20ajox
JXHZRQHZVYDRGX-UHFFFAOYSA-M
sodium;hydrogen sulfate;hydrate
23673662
Reagent
Hygroscopic
Sodium hydrogen sulfate monohydrate
10034-88-5
Crystals
MFCD00149210
Soluble in water. Insoluble in ammonia.
O.[Na+].OS([O-])(=O)=O
138.07
138.07 (120.06anhydrous)
Odorless
White crystalline
Videos
Safety and Handling

Safety and Handling

P261-P271-P280-P305+P351+P338-P310-P501c

H318

missing translation for 'dotInformation' : Hazard Class: 8, Packing Group: III

missing translation for 'einecsNumber' : 231-665-7

missing translation for 'rtecsNumber' : VZ1860000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Product Identifier
  • Sodium hydrogen sulfate monohydrate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Sodium hydrogen sulfate, monohydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only