missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydrogen sulfide, anhydrous, Thermo Scientific Chemicals

Product Code. 11314989
Click to view available options
Quantity:
2 g
10 g

For Use in Paper Pulping, in Dyestuffs Processing, in Rayon and Cellophane Desulfurizing

In paper pulping, in dyestuffs processing, in rayon and cellophane desulfurizing, in unhairing hides and as a bleaching reagent Sodium hydrogen sulfide is used in cloth, paper, pulp and dyestuff industries. It acts as a floating agent in copper mining as well as to remove hairs from hides in leather industries. It is also used as an intermediate in synthetic chemistry. It is utilized as a raw material in the production of ammonium sulphate and ethanethiol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Moisture Sensitive and Hygroscopic
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16721-80-5
Molecular Formula HNaS
Molecular Weight (g/mol) 56.06
MDL Number MFCD00011124
InChI Key HYHCSLBZRBJJCH-UHFFFAOYSA-M
Synonym sodium hydrosulfide, sodium bisulfide, sodium sulfhydrate, sodium hydrogen sulfide, sodium sulfide na sh, sodium sulfanide, sodium mercaptan, sodium mercaptide, sodium hydrosulphide, nahs
PubChem CID 28015
SMILES [Na+].[SH-]

Specifications

Melting Point 350°C
Color White
Physical Form Crystalline
Quantity 2 g
Solubility Information Soluble in water,ether and alcohol.
IUPAC Name sodium sulfanide
Formula Weight 56.06
Odor Like rotten eggs
Sensitivity Hygroscopic; Moisture sensitive
Density 1.79 g/cm3 at 20°C
Chemical Name or Material Sodium hydrogen sulfide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'