missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, 0.1 N standard solution, Thermo Scientific Chemicals

Sodium hydroxide, HNaO, CAS Number-1310-73-2, 7732-18-5, aetznatron, ascarite, caustic soda, soda, caustic, soda lye, sodium hydrate, sodium hydroxide, sodium hydroxide na oh, sodium hydroxide solution, white caustic, 1L, CHEBI:32145, Beige to White, 0.0995 to 0.1005N (20 deg.C), 1.0000g/mL

Brand:  Thermo Scientific Acros 124190025

 View more versions of this product

Product Code. 10348573

  • £38.39 / 2.50 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Volumetric standard solution
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sodium hydroxide
0.1 N Standard Solution
HNaO
0.0995 to 0.1005N (20°C)
01,1083; 05,616; 07,336; 08,460
15, 8761
Solubility in water: soluble. Other solubilities: soluble in alcohol and glycerol
2.5 L
sodium;hydroxide
14798
40
Plastic Bottle
Crystalline Powder
White to Beige
1310-73-2, 7732-18-5
NaOH
MFCD00003548
1.0000g/mL
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
[OH-].[Na+]
39.997
CHEBI:32145
Pure
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only