Learn More
Sodium hydroxide, 0.1 N standard solution, Thermo Scientific Chemicals
Sodium hydroxide, HNaO, CAS Number-1310-73-2, 7732-18-5, aetznatron, ascarite, caustic soda, soda, caustic, soda lye, sodium hydrate, sodium hydroxide, sodium hydroxide na oh, sodium hydroxide solution, white caustic, 1L, CHEBI:32145, Beige to White, 0.0995 to 0.1005N (20 deg.C), 1.0000g/mL
Brand: Thermo Scientific Acros 124190010
17.63 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.
24.16 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Volumetric standard solution
Specifications
Sodium hydroxide | |
0.1 N Standard Solution | |
HNaO | |
0.0995 to 0.1005N (20°C) | |
01,1083; 05,616; 07,336; 08,460 | |
15, 8761 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
Pure | |
Crystalline Powder |
Beige to White | |
1310-73-2, 7732-18-5 | |
NaOH | |
MFCD00003548 | |
1.0000g/mL | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
1 L | |
sodium;hydroxide | |
14798 | |
40 | |
Plastic bottle |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only