missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 0.1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15578344
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Quantity 1 L
Formula Weight 40
Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Poly Bottle
Specific Gravity 1L = 1.00kg
Chemical Name or Material Sodium Hydroxide, 0.1N Volumetric Solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'