missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydroxide, 0.2 N standard solution, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 349685000
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Volumetric standard solutionSpecifications
Sodium hydroxide | |
1310-73-2, 7732-18-5 | |
NaOH | |
MFCD00003548 | |
1.0100g/mL | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
Plastic bottle | |
Liquid |
Colorless | |
HNaO | |
0.1996 to 0.2004N | |
01,1083; 05,616; 07,336; 08,460 | |
15, 8761 | |
Solubility in water: soluble. | |
500 mL | |
sodium;hydroxide | |
14798 | |
39.99 | |
1.01 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Sodium hydroxide, 0.2 N standard solution
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction