missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydroxide, 1.0N Standardized Solution, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
4 L
Description
Sodium hydroxide, 1.0N standardized solution is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 1 L |
Solubility Information | Miscible with glycerol,ethanol and methanol. Immiscible with acetone and ether. |
Formula Weight | 40 |
Appearance | Colourless liquid |
Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
Chemical Name or Material | Sodium hydroxide |
Safety and Handling
Product Identifier
- Sodium hydroxide, 1.0N Standardized Solution
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : sodium hydroxide
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction