missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 2.0N Solution, Water for Injection Quality Water, Biotech Reagent, J.T. Baker™
1681.50 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
19 L
Packaging:
Poly Bottle
Specifications
CAS | 1310-73-2 |
Molecular Formula | HNaO |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Quantity | 19 L |
SMILES | [OH-].[Na+] |
IUPAC Name | sodium;hydroxide |
Molecular Weight (g/mol) | 39.997 |
PubChem CID | 14798 |
ChEBI | CHEBI:32145 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'