missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 32% Solution w/w, for Nitrogen Determination, Fisher Chemical™

Product Code. 16254331
Click to view available options
Quantity:
1 L
5 L

Sodium hydroxide solution suitable for use as a titrant for the determination of nitrogen by Kjeldahl method.

TRUSTED_SUSTAINABILITY

Specifications

Assay Percent Range 32% (w/w)
Quantity 1 L
Packaging HDPE Plastic Bottle
Physical Form Liquid
compliance-icons
Product Identifier
  • 32% w/w Sodium hydroxide solution
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.