missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 32% Solution w/w, for Nitrogen Determination, Fisher Chemical™
Click to view available options
Quantity:
1 L
5 L
Specifications
Assay Percent Range | 32% (w/w) |
Quantity | 5 L |
Packaging | HDPE Plastic Bottle |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'