missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 32% Solution w/w, for Nitrogen Determination, Fisher Chemical™

Sodium hydroxide solution suitable for use as a titrant for the determination of nitrogen by Kjeldahl method.

£51.30 - £106.00

Specifications

Assay Percent Range 32% (w/w)
Packaging HDPE Plastic Bottle
Physical Form Liquid

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16254331
View Documents
Fisher Chemical
S/4970/15
1 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16264331
View Documents
Fisher Chemical
S/4970/21
5 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

32% (w/w)
HDPE Plastic Bottle
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 32% w/w Sodium hydroxide solution

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide

SDS
Documents

Documents

Product Certifications