Learn More
Sodium hydroxide, 40% w/v aq. soln., Thermo Scientific Chemicals
Description
Sodium hydroxide, 40% w/v aq. solution is used as a chemical reactant or neutralization agent in various industries like chemical, pharmaceutical, textile, pulp and paper, metal and food industries and soap and detergent manufacturing. It is also used in the manufacture of synthetic fabrics such as rayon and also for manufacturing of food products. It is utilized for pH adjustment and formation of in situ sodium methylate in bioethanol and biodiesel processing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
UN Number | UN1824 |
Concentration | 40% w/v aq. soln. |
Solubility Information | Miscible with glycerol,ethanol and methanol. Immiscible with acetone and ether. |
Formula Weight | 40 |
Quantity | 4 L |
Physical Form | Liquid |
Chemical Name or Material | Sodium hydroxide |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
X23K500 | Certificate of Analysis | 19/11/2024 | 035639.K2,035639.M4,035639.K7 |
N25K505 | Certificate of Analysis | 04/04/2024 | 035639.K2,035639.M4,035639.K7 |
N07K519 | Certificate of Analysis | 19/03/2024 | 035639.K2,035639.M4,035639.K7 |
N07K518 | Certificate of Analysis | 19/03/2024 | 035639.K2,035639.M4,035639.K7 |
U16J511 | Certificate of Analysis | 24/10/2023 | 035639.K2,035639.M4,035639.K7 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.