missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, 50% w/w aq. soln., Thermo Scientific Chemicals

Product Code. 11347708
Click to view available options
Quantity:
100 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Sodium hydroxide, 50% w/w aq. solution is used as a chemical reactant or neutralization agent in various industries like chemical, pharmaceutical, textile, pulp and paper, metal and food industries and soap and detergent manufacturing. It is also used in the manufacture of synthetic fabrics such as rayon and also for manufacturing food products. It is utilized for pH adjustment and formation of in situ sodium methylate in bioethanol and biodiesel processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color Colorless
Physical Form Liquid
Quantity 100 g
UN Number UN1824
Solubility Information Miscible with glycerol,ethanol and methanol. Immiscible with acetone and ether.
Formula Weight 40
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Density 1.53
Chemical Name or Material Sodium hydroxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'