Learn More
Sodium hydroxide, beads/pellets, 99.99% (metals basis), Thermo Scientific Chemicals
£24.72 - £86.03
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15467065
|
Thermo Scientific Alfa Aesar
045780.06 |
5 g |
£24.72
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15477065
|
Thermo Scientific Alfa Aesar
045780.14 |
25 g |
£37.79
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15487065
|
Thermo Scientific Alfa Aesar
045780.22 |
100 g |
£86.03
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium hydroxide is widely used in a variety of large scale applications such as pulp and paper manufacturing, in the production of soap and detergents as well as water treatment. It finds application in the base- driven hydrolysis of esters, amides and alkyl halides. It is also used as an industrial cleaning agent, an additive in drilling mud to increase alkalinity in bentonite in petroleum industry and as a powerful degreaser on stainless steel and glass bakeware. It is utilized in the Maxam-Gilbert DNA sequencing technique, chocolate and cocoa processing, soft drink processing, caramel coloring production and for thickening ice cream. It is often used to neutralize acids and prepare sodium salts of reagents. It forms a solution with sodium dodecyl sulfate and plays an important role for the decontamination of mycobacteria.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1310-73-2 | |
MFCD00003548 |
Specifications
White | |
316°C | |
2.13 | |
40 | |
3mm Hg at 800°C |
1310-73-2 | |
MFCD00003548 | |
Soluble in water,methanol,glycerol and ethanol. Insoluble in acetone and ether. | |
Odorless | |
Sodium hydroxide |
Safety and Handling
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 215-185-5
missing translation for 'rtecsNumber' : WB4900000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Danger
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.