missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, Certified AR for Analysis, 30% Solution, d=1.33, Fisher Chemical™
£25.35 - £82.25
Chemical Identifiers
CAS | 1310-73-2 |
---|---|
Molecular Formula | HNaO |
MDL Number | 3548 |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
PubChem CID | 14798 |
ChEBI | CHEBI:32145 |
SMILES | [OH-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10060622
|
Fisher Chemical
S/4950/PB15 |
1 L |
£25.35
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10263912
|
Fisher Chemical
S/4950/PB17 |
2.5 L |
£29.95
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15960957
|
Fisher Chemical
S/4950/21 |
5 L |
£82.25
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifications
12 | |
120°C | |
8°C | |
3548 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
HDPE Plastic Bottle |
Colorless | |
1310-73-2 | |
HNaO | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sodium hydroxide, 30% solution, d=1,33
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Sodium hydroxide