missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, Certified AR for Analysis, 46-51% Solution, Certified AR, Fisher Chemical™
£36.35 - £655.00
Chemical Identifiers
CAS | 1310-73-2 |
---|---|
Molecular Formula | HNaO |
Molecular Weight (g/mol) | 39.997 |
MDL Number | 3548 |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
PubChem CID | 14798 |
ChEBI | CHEBI:32145 |
IUPAC Name | sodium;hydroxide |
SMILES | [OH-].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10502343
|
Fisher Chemical
S/4930/05 |
100 mL | HDPE plastic bottle |
£36.35
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10010650
|
Fisher Chemical
S/4930/15 |
1 L | HDPE plastic bottle |
£49.35
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10795721
|
Fisher Chemical
S/4930/17 |
2.5 L | HDPE plastic bottle |
£95.35
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10366290
|
Fisher Chemical
S/4930/21 |
5 L | HDPE plastic bottle |
£157.00
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10376290
|
Fisher Chemical
S/4930/25 |
25 L | Plastic container |
£655.00
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
1310-73-2 | |
39.997 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
HNaO | |
3548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
Specifications
13 | |
1310-73-2 | |
HNaO | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
14mmHg |
145°C | |
12°C | |
3548 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
> 1.0 | |
Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sodium hydroxide, 50 wt% solution in water
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Sodium hydroxide