missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, Certified AR for Analysis, Pellets, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Product Code. 10254540 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
1 kg
3 kg
5 kg
Unit Size:
1kg
3kg
500g
5kg
This item is not returnable. View return policy

Product Code. 10254540

Brand: Fisher Chemical S/4920/63

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HNaO, CAS Number-1310-73-2, aetznatron, ascarite, caustic soda, soda, caustic, soda lye, sodium hydrate, sodium hydroxide, sodium hydroxide na oh, sodium hydroxide solution, white caustic, 1kg, 1390 deg.C, CHEBI:32145, White, 40g/mol, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 318 deg.C, 3548 | CAS: 1310-73-2 | HNaO | 39.997 g/mol

HNaO, CAS Number-1310-73-2, white caustic, aetznatron, sodium hydroxide na oh, caustic soda, ascarite, soda lye, soda, caustic, sodium hydroxide solution, sodium hydrate, sodium hydroxide, 5kg, 1390 deg.C, CHEBI:32145, White, 40g/mol, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 318 deg.C, 3548

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number 3548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

pH 14
Color White
Boiling Point 1390°C
Melting Point 318°C
Quantity 3 kg
Formula Weight 40g/mol
Packaging Plastic powder jar
Vapor Pressure 1mbar 700
Physical Form Solid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.