missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, DILUT-IT Concentrate, 0.5N, J.T. Baker™
Click to view available options
Quantity:
For dilution to 1 L
Packaging:
LDPE ampule
Description
DILUT-IT solutions are relied upon for fast, accurate results in life sciences and pharmaceutical quality control applications. Simply dilute the volumetric concentrate to 1 liter (carefully following instructions provided with the package) and the resulting solution will have an exact normality within ±0.1%.
- Enhanced productivity: Eliminate expensive, time-consuming steps, such as the selection and analysis of starting reagents, and weighing and standardization of the final solution
- Space saving: Products require only one-tenth of the space required for ready-made solutions
- Reduced shipping costs: Spend less on shipping due to the reduced weight and package size of a concentrate
Specifications
Specifications
Color | Colorless |
Boiling Point | 100°C |
CAS | 1310-73-2 |
Melting Point | 0°C |
Molecular Formula | HNaO |
Concentration | 0.5mol/L |
Density | 1.2g/cm3 at 20°C |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
Solubility Information | Fully miscible in water |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Show More |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Hydroxide, DILUT-IT Concentrate, 0.5N, J.T. Baker™ >
Spot an opportunity for improvement?Share a Content Correction