missing translation for 'onlineSavingsMsg'
Learn More
Sodium Hydroxide Ethanolic Solution, Fisher Chemical™ Product Code.: 12930894
compliance-icons

Sodium Hydroxide Ethanolic Solution, Fisher Chemical™
BIOPHARMA

Product Code. 12930894
1000 mL
Click to view available options
Quantity:
1000 mL
This item is not returnable. View return policy

Product Code. 12930894

Brand: Fisher Chemical VL 055 1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Quantity 1000 mL
Physical Form Liquid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1101757 Certificate TypeCertificate of Analysis Date02/04/2025 Catalog NumberVL 055 1000
Lot Number1079396 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberVL 055 1000
Lot Number1051084 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberVL 055 1000
Lot Number1013893 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberVL 055 1000
compliance-icons
Product Identifier
  • Sodium hydroxide 0.1M Ethanolic. European Pharmacopoeia Ref: 3007000 (VL 055)
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Hydroxide Ethanolic Solution, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.