missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, Extra Pure, SLR, Pearl, Fisher Chemical™
41.57 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
1 kg
3 kg
Specifications
pH | 14 |
Color | White |
Boiling Point | 1390°C |
Melting Point | 318°C |
Quantity | 1 kg |
Formula Weight | 40g/mol |
Packaging | Plastic powder jar |
Vapor Pressure | 1mbar 700 |
Physical Form | Solid |
Safety and Handling
Product Identifier
- Sodium hydroxide
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'