missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, Extra Pure, SLR, Pellets, Fisher Chemical™

£38.10 - £225.00

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number 3548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10388103
View Documents Promotion Details
Fisher Chemical
S/4880/50
250 g
£40.70
250g
Please to purchase this item. Need a web account? Register with us today!
10478030
View accessory products View Documents Promotion Details
Fisher Chemical
S/4880/53
500 g
£38.10
500g
Please to purchase this item. Need a web account? Register with us today!
10306200
View accessory products View Documents Promotion Details
Fisher Chemical
S/4880/60
1 kg
£54.45
1kg
Please to purchase this item. Need a web account? Register with us today!
10665502
View Documents
Fisher Chemical
S/4880/63
3 kg
£156.00
3kg
Please to purchase this item. Need a web account? Register with us today!
10090632
View Documents
Fisher Chemical
S/4880/65
5 kg
£225.00
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

1310-73-2
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
HNaO
3548
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]

Specifications

14
1390°C
318°C
3548
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40g/mol
1mbar 700
White
1310-73-2
HNaO
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
[OH-].[Na+]
39.997
CHEBI:32145
Plastic powder jar
Solid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sodium hydroxide

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications