missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, flake, 98%

Product Code. 11438327
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
This item is not returnable. View return policy

Product Code. 11438327

Brand: Thermo Scientific Alfa Aesar A18395.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1310-73-2 | HNaO | 39.997 g/mol

Sodium hydroxide flake is widely used in a variety of large scale applications such as pulp and paper manufacturing, in the production of soap and detergents as well as in water treatment plants. It is used in the Bayer process of aluminum manufacture as well as to remove sulfurous impurities from crude oil. It is also used in the manufacture of biodiesel, rayon and cellophane; as a catalyst for the trans esterification of methanol and triglycerides and as paint stripper on wooden objects. It is mainly used for pH control, acid neutralization, off-gas scrubbing and as a catalyst in various industries like pharmaceutical, agrochemical, dyes, paints and polymer. It is also used in refining vegetable oils, rubber reclamation, metal processing, metal degreasing, adhesive preparations, paint removers, disinfectants, rubber latex stabilizers and in the stabilization of sodium hypochlorite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hydroxide flake is widely used in a variety of large scale applications such as pulp and paper manufacturing, in the production of soap and detergents as well as in water treatment plants. It is used in the Bayer process of aluminum manufacture as well as to remove sulfurous impurities from crude oil. It is also used in the manufacture of biodiesel, rayon and cellophane; as a catalyst for the trans esterification of methanol and triglycerides and as paint stripper on wooden objects. It is mainly used for pH control, acid neutralization, off-gas scrubbing and as a catalyst in various industries like pharmaceutical, agrochemical, dyes, paints and polymer. It is also used in refining vegetable oils, rubber reclamation, metal processing, metal degreasing, adhesive preparations, paint removers, disinfectants, rubber latex stabilizers and in the stabilization of sodium hypochlorite.

Notes
Air sensitive and hygroscopic. It is deliquescent and readily absorbs moisture and carbon dioxide in air. Incompatible with strong oxidizing agents, strong acids and organic materials.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

pH 14
Boiling Point 1390°C
Melting Point 318°C
UN Number UN1823
Density 2.13
Merck Index 14,8627
Solubility Information Soluble in water,methanol,glycerol and ethanol. Insoluble in acetone and ether.
Quantity 500 g
Formula Weight 40
Percent Purity 98%
Odor Odorless
Physical Form Flakes
Chemical Name or Material Sodium hydroxide
Show More Show Less
compliance-icons
Product Identifier
  • Sodium hydroxide, flake
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.