Learn More
Sodium hydroxide (low chloride), ACS, 97.0% min, Thermo Scientific Chemicals
Description
In chemical manufacture, rayon and cellophane, as a neutralizing agent in petroleum refining, in pulp, paper detergents and textiles, and as a lab reagent. NaOH solutions precipitate most metals as hydroxides from aqueous solutions of the metal saltsSodium hydroxide is widely used in variety of large scale applications such as pulp and paper manufacturing, in the production of soap and detergents as well as water treatment. It finds application in the base- driven hydrolysis of esters, amides and alkyl halides. It is also used as an industrial cleaning agent, an additive in drilling mud to increase alkalinity in bentonite in petroleum industry and as a powerful degreaser on stainless steel and glass bakeware. It is utilized in the Maxam-Gilbert DNA sequencing technique, chocolate and cocoa processing, soft drink processing, caramel coloring production and for thickening ice cream. It is often used to neutralize acids and prepare sodium salts of reagents. It forms a solution with sodium dodecyl sulfate and plays an important role for the decontamination of mycobacteria.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 1390°C |
Melting Point | 318°C |
Color | White |
Physical Form | Pellets |
Quantity | 10 kg |
Assay Percent Range | 97.0% min. |
Solubility Information | Soluble in water,alcohol and glycerolSoluble in water,glycerol,ethanol and methanol. Insoluble in acetone and ether. |
Formula Weight | 40 |
Grade | ACS |
Odor | Odorless |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M03L014 | Certificate of Analysis | 21/01/2025 | 013455.36,013455.A3,013455.22,013455.F6,013455.A9 |
X07K032 | Certificate of Analysis | 15/10/2024 | 013455.36,013455.A3,013455.22,013455.F6,013455.A9 |
W03K003 | Certificate of Analysis | 30/09/2024 | 013455.36,013455.A3,013455.22,013455.F6,013455.A9 |
P23H020 | Certificate of Analysis | 05/03/2024 | 013455.36,013455.A3,013455.22,013455.F6,013455.A9 |
Z28J027 | Certificate of Analysis | 09/01/2024 | 013455.36,013455.A3,013455.22,013455.F6,013455.A9 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.