missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, pure, 1N Standard Solution, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 124260250


 View more versions of this product

Product Code. 11974131

  • £219.09 / 25 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Volumetric standard solution
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sodium hydroxide
1310-73-2, 7732-18-5
NaOH
01,1083; 05,616; 07,336; 08,460
15, 8761
Solubility in water: soluble. Other solubilities: soluble in alcohol and glycerol
25 L
sodium;hydroxide
14798
40
1.04
Colorless
HNaO
0.998 to 1.002N (20°C)
1.0400g/mL
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
[OH-].[Na+]
39.997
CHEBI:32145
Plastic Can
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sodium hydroxide, 1N standard solution

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only