missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, Pure, Solution 2% (v/v), Fisher Chemical

Product Code. 12867182
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Color Colorless
Boiling Point 100°C
CAS 1310-73-2
Molecular Formula HNaO
MDL Number 3548
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 500 mL
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Packaging HDPE plastic bottle
Vapor Pressure 23 &20
Physical Form Liquid
Mehr anzeigen Weniger anzeigen
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'