missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, purum p.a., ≥98% (T), ACS Reagent, Honeywell™
Purum p.a.,≥98.0% (T), beads (small)
£33.43 - £163.42
Chemical Identifiers
CAS | 1310-73-2 |
---|---|
Molecular Formula | HNaO |
Molecular Weight (g/mol) | 39.997 |
MDL Number | MFCD00003548 |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
PubChem CID | 14798 |
ChEBI | CHEBI:32145 |
IUPAC Name | sodium;hydroxide |
SMILES | [OH-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15676820
|
Honeywell Research Chemicals
71691-250G |
250 g | N/A | ||||||
15666820
|
Honeywell Research Chemicals
71691-1KG |
1 kg | N/A | ||||||
15686820
|
Honeywell Research Chemicals
71691-5KG |
5 kg | N/A | ||||||
15696820
|
Honeywell Research Chemicals
71691-6X1KG |
6 x 1 kg | N/A | ||||||
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Chemical Identifiers
1310-73-2 | |
39.997 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
HNaO | |
MFCD00003548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
Specifications
1310-73-2 | |
NaOH | |
UN1823 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
40g/mol | |
>1 (vs air) | |
Sodium hydroxide |
HNaO | |
MFCD00003548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
≥98% (T) | |
3 mmHg (37°C), 18 mmHg (20°C) |
Safety and Handling
P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
missing translation for 'rtecsNumber' : WB4900000