missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, Semiconductor grade, Honeywell
Pellets, semiconductor grade, 99.99% trace metals basis
£47.37 - £450.60
Chemical Identifiers
CAS | 1310-73-2 |
---|---|
Molecular Formula | HNaO |
Molecular Weight (g/mol) | 39.997 |
MDL Number | MFCD00003548 |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
PubChem CID | 14798 |
ChEBI | CHEBI:32145 |
IUPAC Name | sodium;hydroxide |
SMILES | [OH-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15683080
|
Honeywell Research Chemicals
306576-25G |
25 g |
£47.37
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15673080
|
Honeywell Research Chemicals
306576-100G |
100 g |
£126.85
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15693080
|
Honeywell Research Chemicals
306576-500G |
500 g |
£450.60
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Comprehensive portfolio for the electronics and semiconductor industry
- For scientists engaged in education, research, development, prototyping and small scale fabrication of high tech micro- and nanochips
- Superior purity, stability and exceptional lot-to-lot consistency
- Includes high purity products required for high performance applications such as etchants, solvents or hexamethyldisilazane (HMDS)
Chemical Identifiers
1310-73-2 | |
39.997 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
HNaO | |
MFCD00003548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
Specifications
1310-73-2 | |
NaOH | |
1823 | |
Soluble in water, 1260 g/L at 20°C | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
99.99% (trace metals basis) | |
>1 (vs air) | |
Sodium Hydroxide |
HNaO | |
MFCD00003548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
40g/mol | |
Semiconductor | |
18mmHg (20°C), 3mmHg (37°C) |
Safety and Handling
P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H290-H314
missing translation for 'einecsNumber' : 215-185-5
missing translation for 'rtecsNumber' : WB4900000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sodium hydroxide
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Sodium hydroxide