missing translation for 'onlineSavingsMsg'
Learn More
Sodium Hydroxide Solution 0.1M (0.1N), NIST Standard Solution ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™ Product Code.: 10637032

Sodium Hydroxide Solution 0.1M (0.1N), NIST Standard Solution ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Product Code. 10637032
5 L, HDPE plastic bottle
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
Packaging:
Bag in box
HDPE plastic bottle
This item is not returnable. View return policy

Product Code. 10637032

Brand: Fisher Chemical J/7660/21

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

HNaO, CAS Number-1310-73-2, aetznatron, ascarite, caustic soda, soda, caustic, soda lye, sodium hydrate, sodium hydroxide, sodium hydroxide na oh, sodium hydroxide solution, white caustic, 1L, 100 deg.C, CHEBI:32145, Colorless, HEMHJVSKTPXQMS-UHFFFAOYSA-M, sodium;hydroxide, 3548, 39.997

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number 3548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, causticShow MoreShow Less
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color Colorless
Boiling Point 100°C
Melting Point 0°C
Quantity 5 L
Packaging HDPE plastic bottle
Vapor Density > 1.0
Physical Form Liquid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2525697 Certificate TypeCertificate of Analysis Date07/02/2025 Catalog NumberJ/7660C/90,J/7660/24,J/7660C/05,J/7660/21,J/7660/17,J/7660/15
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155

    Expires2025-04-30

    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions See Full Promo Details
  • Multiple Manufacturers
    Quality testing products for all your needs.
    Promo Code: 24134

    Expires2025-03-31

  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Hydroxide Solution 0.1M (0.1N), NIST Standard Solution ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.