missing translation for 'onlineSavingsMsg'
Learn More
Sodium Hydroxide, 0.1M, EP Grade, Reagecon™ Product Code.: 11484809

Sodium Hydroxide, 0.1M, EP Grade, Reagecon™
BIOPHARMA

Product Code. 11484809 Shop All Reagecon Products
1 L, Tamper evident
Click to view available options
Quantity:
1 L
Packaging:
Tamper evident
This item is not returnable. View return policy

Product Code. 11484809

Brand: Reagecon 3006600

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Manufactured according to European Pharmacopoeia Chapter 4 (4.2.2) and prepared in an ISO 9001 facility. Sodium Hydroxide, 0.1M, EP Grade, Reagecon™ is a ready-to-use volumetric solution with 3.9997g/L NaOH in water for use in manual and automated titrations.

Sodium Hydroxide is an inorganic compound consisting of sodium cations Na+ and hydroxide anions OH. It is frequently used a matrix and reagent in a variety of laboratory applications.

This solution is prepared by diluting 100mL of 1M sodium hydroxide solution to 1L with CO2-free water (where 1M sodium hydroxide solution contains 42g NaOH diluted to 1L with CO2-free water).
  • Manufactured in an ISO 9001-compliant facility according to Eu. Ph. Chapter 4 (4.2.2) guidelines
  • Provided with a certificate of analysis and a label that shows the lot and expiration date
  • Ready to use product eliminates the time and labor of preparing precise volumetric solutions

Specifications

Color Colorless
CAS 1310-73-2, 7732-18-5
Molecular Formula HNaO
Concentration 0.1M = 3.9997g/L
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Solubility Information Soluble in water
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 1 L
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 39.997g/mol
Grade Eu. Ph. Chapter 4(4.2.2)
Packaging Tamper evident
Solvent or Matrix Water
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS20125C2 Certificate TypeCertificate of Analysis Date08/04/2025 Catalog Number3006600
Lot NumberS20124L1 Certificate TypeCertificate of Analysis Date26/03/2025 Catalog Number3006600
Lot NumberS20124M1 Certificate TypeCertificate of Analysis Date16/03/2025 Catalog Number3006600
Lot NumberS20124J2 Certificate TypeCertificate of Analysis Date07/01/2025 Catalog Number3006600
Lot NumberS20124G1 Certificate TypeCertificate of Analysis Date01/09/2024 Catalog Number3006600
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Hydroxide, 0.1M, EP Grade, Reagecon™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.